Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145781.2 | NP_665724.1 | 1, 2 | 165 | ||
GenBank mRNA | AF144701.1 | - | 1, 2 | 165 |