Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148892.3 | NP_683690.2 | 4 | 327 | ||
XM_008760287.1 | XP_008758509.1 | 4 | 322 | |||
GenBank mRNA | AF523266.1 | - | 4 | 206 | ||
BC062400.1 | - | 4 | 306 | 5621572 | ||
FM048871.1 | - | 4 | 327 |