Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144752.2 | NP_653353.2 | 2 | 465 | ||
GenBank mRNA | AF068268.1 | - | 477 | |||
BC091121.1 | - | 2 | 465 | 7383389 |