Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145090.3 | NP_659558.1 | 12 | 1079 | ||
XM_006235731.2 | XP_006235793.1 | 13 | 1209 | |||
XM_006235732.1 | XP_006235794.1 | 13 | 1150 | |||
GenBank mRNA | BC070895.1 | - | 11 | 1520 | 7099840 | |
CV120035.1 | - | 4 | 170 | |||
DQ167811.1 | - | 11 | 826 | |||
FQ222295.1 | - | 12 | 1102 | |||
U35776.1 | - | 12 | 1079 |