Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005387.6 | NP_005378.4 | 18 | 2764 | ||
NM_016320.4 | NP_057404.2 | 18 | 2713 | |||
NM_139131.4 | NP_624357.1 | 18 | 2713 | |||
NM_139132.3 | NP_624358.2 | 18 | 2713 | |||
GenBank mRNA | AB040538.1 | - | 18 | 2519 | ||
AF071076.1 | - | 18 | 2416 | |||
AF071077.1 | - | 18 | 2416 | |||
AF231130.1 | - | 18 | 2436 | |||
AK292882.1 | - | 18 | 2504 | |||
AK301512.1 | - | 18 | 2172 | |||
BC041136.1 | - | 18 | 2462 | 5179730 | ||
U41815.1 | - | 18 | 2436 |