Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145096.2 | NP_659564.2 | 10 | 1154 | ||
GenBank mRNA | AB217870.1 | - | 10 | 953 | ||
AF228917.1 | - | 10 | 1143 | |||
AY886521.1 | - | 10 | 1154 |