Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145677.1 | NP_663710.1 | 2 | 359 | ||
XM_006233931.2 | XP_006233993.1 | 2 | 480 | |||
XM_006233932.2 | XP_006233994.1 | 2 | 480 | |||
XM_006233933.2 | XP_006233995.1 | 2 | 1132 | |||
XM_006233934.2 | XP_006233996.1 | 2 | 1132 | |||
XM_006233935.2 | XP_006233997.1 | 2 | 1696 | |||
XM_006233937.2 | XP_006233999.1 | 2 | 716 | |||
GenBank mRNA | AY043169.1 | - | 2 | 359 | ||
FQ211394.1 | - | 2 | 375 |