Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145678.1 | NP_663711.1 | 8, 9 | 859 | ||
XM_006255560.1 | XP_006255622.1 | 8, 9 | 1078 | |||
GenBank mRNA | AB076398.1 | - | 8, 9 | 859 | ||
BC070931.1 | - | 8, 9 | 962 | 7096867 | ||
FQ213218.1 | - | 1039 |