Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145770.2 | NP_665713.2 | 10, 11 | 1552 | ||
XM_006251743.2 | XP_006251805.1 | 11, 12 | 1637 | |||
XM_006251744.2 | XP_006251806.1 | 10, 11 | 1508 | |||
XM_006251745.2 | XP_006251807.1 | 11, 12 | 1555 | |||
GenBank mRNA | X95189.1 | - | 10, 11 | 1551 |