Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145783.1 | NP_665726.1 | 4 | 392 | ||
GenBank mRNA | FQ210272.1 | - | 4 | 425 | ||
FQ211124.1 | - | 4 | 452 | |||
FQ217304.1 | - | 4 | 425 | |||
FQ217698.1 | - | 4 | 415 | |||
FQ223560.1 | - | 4 | 425 | |||
FQ223883.1 | - | 4 | 409 | |||
FQ224560.1 | - | 4 | 417 | |||
FQ224768.1 | - | 4 | 418 | |||
FQ228531.1 | - | 4 | 425 | |||
X15030.1 | - | 4 | 392 |