Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145789.2 | NP_665732.2 | 5 | 668 | ||
GenBank mRNA | AY044251.1 | - | 5 | 652 | ||
BC093615.1 | - | 5 | 668 | 7310384 |
Gene Symbol: LOC100360218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751077.3 | XP_003751125.1 | 1 | 694 |