Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153467.1 | NP_703197.1 | 3 | 253 | ||
XM_006236545.2 | XP_006236607.1 | 3 | 762 | |||
XM_008762878.1 | XP_008761100.1 | 4 | 689 | |||
GenBank mRNA | AF294688.1 | - | 3 | 253 | ||
FQ213688.1 | - | 3 | 215 |