Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153468.1 | NP_703198.1 | 1, 2 | 78 | ||
GenBank mRNA | AB082125.1 | - | 1, 2 | 78 |