Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153626.1 | NP_705890.1 | 1 | 198 | ||
XM_006230697.2 | XP_006230759.1 | 3 | 2355 | |||
GenBank mRNA | AB050103.1 | - | 1 | 198 |