Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152936.1 | NP_690919.1 | 3 | 192 | ||
GenBank mRNA | FQ209906.1 | - | 3 | 173 | ||
FQ210000.1 | - | 3 | 206 | |||
FQ210029.1 | - | 3 | 205 | |||
FQ210138.1 | - | 3 | 205 | |||
FQ210344.1 | - | 3 | 205 | |||
FQ210442.1 | - | 3 | 205 | |||
FQ210447.1 | - | 3 | 205 | |||
FQ210766.1 | - | 3 | 207 | |||
FQ210836.1 | - | 3 | 207 | |||
FQ210933.1 | - | 3 | 207 | |||
FQ211109.1 | - | 3 | 205 | |||
FQ211214.1 | - | 3 | 209 | |||
FQ211434.1 | - | 3 | 191 | |||
FQ218107.1 | - | 3 | 207 | |||
FQ218122.1 | - | 3 | 206 | |||
FQ218165.1 | - | 3 | 207 | |||
FQ218208.1 | - | 3 | 203 | |||
FQ218422.1 | - | 3 | 205 | |||
M27883.1 | - | 3 | 192 |
Gene Symbol: LOC100911558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751771.3 | XP_003751819.1 | 3 | 542 |