Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153295.1 | NP_695207.1 | 2, 3 | 241 | ||
GenBank mRNA | AF488741.1 | - | 2, 3 | 241 |
Gene Symbol: LOC103693583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768769.1 | XP_008766991.1 | 3, 4 | 935 |