Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153318.1 | NP_695230.1 | 3 | 343 | ||
XM_006241049.1 | XP_006241111.1 | 3 | 417 | |||
XM_008765156.1 | XP_008763378.1 | 3 | 496 | |||
GenBank mRNA | BC078713.1 | - | 3 | 253 | 7107801 | |
U39208.1 | - | 3 | 343 |