Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Soat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153728.2 | NP_714950.2 | 2, 3 | 420 | ||
GenBank mRNA | AB075946.1 | - | 2, 3 | 152 | ||
AB101480.1 | - | 2, 3 | 420 |