Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Canx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172008.2 | NP_742005.1 | 3 | 318 | ||
XM_008767679.1 | XP_008765901.1 | 3 | 439 | |||
XM_008767680.1 | XP_008765902.1 | 3 | 358 | |||
GenBank mRNA | CB586187.1 | - | 3 | 318 | ||
EV776124.1 | - | 1 | 47 | |||
FQ210638.1 | - | 3 | 344 | |||
L18889.1 | - | 2 | 215 |