Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152242.2 | NP_689448.2 | 6, 7 | 1196 | ||
XM_006255066.2 | XP_006255128.1 | 6, 7 | 2248 | |||
XM_008772264.1 | XP_008770486.1 | 6, 7 | 1398 | |||
XM_008772265.1 | XP_008770487.1 | 7, 8 | 1662 | |||
XM_008772266.1 | XP_008770488.1 | 6, 7 | 2908 | |||
XM_008772267.1 | XP_008770489.1 | 6, 7 | 3171 | |||
XM_008772268.1 | XP_008770490.1 | 7, 8 | 1495 | |||
XM_008772269.1 | XP_008770491.1 | 6, 7 | 2277 | |||
XM_008772270.1 | XP_008770492.1 | 6, 7 | 2753 | |||
GenBank mRNA | AF529886.1 | - | 6, 7 | 1197 | ||
CV116515.1 | - | 4, 5 | 652 | |||
EX488169.1 | - | 3, 4 | 307 |
Set Membership: |
G-protein coupled receptor |