Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152846.1 | NP_690059.1 | 8 | 812 | ||
GenBank mRNA | AF532184.1 | - | 8 | 812 |