Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153306.1 | NP_695218.1 | 3 | 586 | ||
GenBank mRNA | AB036990.1 | - | 3 | 586 |