Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153625.1 | NP_705889.1 | 13 | 2063 | ||
XM_006248422.2 | XP_006248484.1 | 14 | 2243 | |||
XM_006248423.2 | XP_006248485.1 | 14 | 2208 | |||
XM_006248424.2 | XP_006248486.1 | 14 | 2492 | |||
XM_006248425.2 | XP_006248487.1 | 14 | 2081 | |||
XM_006248426.2 | XP_006248488.1 | 13 | 2129 | |||
XM_006248427.2 | XP_006248489.1 | 13 | 2096 | |||
XM_006248429.2 | XP_006248491.1 | 12 | 2064 | |||
XM_008768704.1 | XP_008766926.1 | 14 | 2165 | |||
XM_008768706.1 | XP_008766928.1 | 12 | 1986 | |||
GenBank mRNA | AB024430.1 | - | 13 | 2063 | ||
AB024431.1 | - | 11 | 1806 |