Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172010.1 | NP_742007.1 | 4 | 468 | ||
GenBank mRNA | AF508020.1 | - | 4 | 468 | ||
BC088107.1 | - | 4 | 424 | 7368731 |