Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172018.1 | NP_742015.1 | 2 | 262 | ||
XM_006247874.1 | XP_006247936.1 | 2 | 455 | |||
XM_006247875.1 | XP_006247937.1 | 2 | 455 | |||
GenBank mRNA | AF508021.1 | - | 2 | 262 | ||
AY327509.1 | - | 2 | 262 | |||
AY327510.1 | - | 2 | 262 |