Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdlbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172039.2 | NP_742036.2 | 4 | 400 | ||
XM_006245548.1 | XP_006245610.1 | 4 | 440 | |||
GenBank mRNA | BC063147.1 | - | 4 | 400 | ||
BC105748.1 | - | 4 | 400 | 5598374 | ||
U90725.1 | - | 4 | 400 |