Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172327.2 | NP_758830.2 | 1 | 80 | ||
XM_006230699.2 | XP_006230761.1 | 2 | 278 | |||
XM_006230700.2 | XP_006230762.1 | 2 | 247 | |||
GenBank mRNA | AF543560.1 | - | 1 | 114 | ||
BC089110.1 | - | 1 | 80 | 7311852 | ||
FQ214270.1 | - | 1 | 188 | |||
FQ228146.1 | - | 1 | 108 |