Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172336.3 | NP_758839.3 | 1, 2 | 203 | ||
XM_006228980.1 | XP_006229042.1 | 2, 3 | 481 | |||
GenBank mRNA | AY123225.1 | - | 1, 2 | 203 | ||
BC061786.1 | - | 2, 3 | 481 | 5623784 |