Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173143.1 | NP_775166.1 | 2 | 220 | ||
GenBank mRNA | BC097273.1 | - | 278 | |||
FQ222350.1 | - | 590 | ||||
U94904.1 | - | 2 | 220 |