Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175578.3 | NP_783168.1 | 2, 3 | 461 | ||
XM_006244593.2 | XP_006244655.1 | 1, 2 | 385 | |||
GenBank mRNA | AF459022.1 | - | 1, 2 | 183 | ||
AF459023.1 | - | 2, 3 | 303 | |||
BC086974.1 | - | 2, 3 | 461 | 7193921 |