Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018490.2 | NP_060960.2 | 3 | 790 | ||
GenBank mRNA | AB209743.1 | - | 3 | 709 | ||
AF257182.1 | - | 3 | 749 | |||
BC033039.2 | - | 2 | 706 | 5260923 |