Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172075.2 | NP_742072.2 | 3 | 442 | ||
XM_006240995.2 | XP_006241057.1 | 3 | 462 | |||
GenBank mRNA | BC081706.1 | - | 3 | 369 | 7111806 | |
M61142.1 | - | 3 | 365 |