Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145846.2 | NP_001139318.1 | 4 | 279 | ||
NM_172077.2 | NP_742074.2 | 4 | 381 | |||
GenBank mRNA | CF249035.1 | - | 3 | 268 | ||
D23676.1 | - | 4 | 279 | |||
L10229.1 | - | 4 | 381 |