Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172091.2 | NP_742088.1 | 14 | 1406 | ||
NM_172092.2 | NP_742089.1 | 14 | 1414 | |||
XM_006247876.2 | XP_006247938.1 | 14 | 1520 | |||
XM_006247877.1 | XP_006247939.1 | 12 | 1245 | |||
XM_008768468.1 | XP_008766690.1 | 13 | 1463 | |||
XM_008768469.1 | XP_008766691.1 | 13 | 1680 | |||
GenBank mRNA | BC081700.1 | - | 13 | 1271 | 7192550 | |
L04796.1 | - | 14 | 1376 | |||
M96674.1 | - | 14 | 1416 | |||
X68692.1 | - | 14 | 1376 |
Set Membership: |
G-protein coupled receptor |