Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172322.1 | NP_758825.1 | 3 | 432 | ||
GenBank mRNA | AB053165.1 | - | 3 | 432 | ||
FQ226280.1 | - | 3 | 590 | |||
FQ227544.1 | - | 3 | 612 |