Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172327.2 | NP_758830.2 | 4 | 532 | ||
XM_006230699.2 | XP_006230761.1 | 5 | 730 | |||
XM_006230700.2 | XP_006230762.1 | 5 | 699 | |||
GenBank mRNA | AF543560.1 | - | 4 | 566 | ||
BC089110.1 | - | 4 | 532 | 7311852 | ||
FQ214270.1 | - | 4 | 640 | |||
FQ228146.1 | - | 4 | 560 |