Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172328.2 | NP_758831.1 | 4, 5 | 300 | ||
GenBank mRNA | AF521561.1 | - | 4, 5 | 300 | ||
AY471575.1 | - | 4, 5 | 300 |