Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175577.3 | NP_783167.2 | 5 | 909 | ||
XM_006231021.2 | XP_006231083.1 | 5 | 1016 | |||
XM_008760218.1 | XP_008758440.1 | 5 | 1016 | |||
GenBank mRNA | AF488727.1 | - | 5 | 848 | ||
BC089767.1 | - | 5 | 909 | 7383438 |
Gene Symbol: LOC100911366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760255.1 | XP_008758477.1 | 4 | 946 |