Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172021.2 | NP_742018.2 | 8 | 1082 | ||
XM_006247266.2 | XP_006247328.1 | 7 | 2229 | |||
XM_006247267.2 | XP_006247329.1 | 5 | 1370 | |||
XM_008767953.1 | XP_008766175.1 | 7 | 2232 | |||
GenBank mRNA | AJ278800.1 | - | 7 | 1062 | ||
BC076503.1 | - | 8 | 1082 | 7100584 |