Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172034.2 | NP_742031.1 | 2 | 224 | ||
GenBank mRNA | BC087675.1 | - | 2 | 224 | 7319293 | |
M69056.1 | - | 2 | 238 |