Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172047.1 | NP_742044.1 | 2 | 302 | ||
XM_008768707.1 | XP_008766929.1 | 2 | 189 | |||
GenBank mRNA | AY049022.1 | - | 2 | 302 |