Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018980.2 | NP_061853.1 | 1 | 515 | ||
GenBank mRNA | BC069509.1 | - | 1 | 370 | 7262216 | |
BC095522.1 | - | 1 | 515 | 30706589 | ||
BC101586.1 | - | 1 | 391 | 8069092 | ||
BC101588.1 | - | 1 | 391 | 8069094 |