Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034023.1 | NP_001029195.1 | 2 | 212 | ||
NM_001034024.1 | NP_001029196.1 | 2 | 172 | |||
NM_019100.4 | NP_061973.1 | 1 | 393 | |||
XM_005271037.4 | XP_005271094.1 | 1 | 383 | |||
XM_005271039.1 | XP_005271096.1 | 2 | 283 | |||
XM_005271040.1 | XP_005271097.1 | 2 | 160 | |||
XM_011541784.2 | XP_011540086.1 | 2 | 217 | |||
XM_011541785.2 | XP_011540087.1 | 2 | 237 | |||
XM_017001807.1 | XP_016857296.1 | 2 | 268 | |||
XM_017001808.1 | XP_016857297.1 | 2 | 271 | |||
GenBank mRNA | AB037846.1 | - | 2 | 177 | ||
AF265228.1 | - | 1 | 92 | |||
AK021605.1 | - | 2 | 156 | |||
AK289366.1 | - | 2 | 195 | |||
AK293598.1 | - | 2 | 203 | |||
AK298793.1 | - | 2 | 185 | |||
AK300217.1 | - | 2 | 170 | |||
AK300372.1 | - | 2 | 162 | |||
AK300442.1 | - | 2 | 200 | |||
AL136657.1 | - | 2 | 174 | |||
AL137200.1 | - | 2 | 176 | |||
BC002855.2 | - | 2 | 161 | 3635724 | ||
BC008053.2 | - | 1 | 355 | 3834056 | ||
BM827551.1 | - | 2 | 224 | |||
CA842195.1 | - | 2 | 204 | |||
CB123593.1 | - | 1 | 382 |