Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238385.2 | XP_006238447.1 | 9 | 1291 | ||
XM_006238388.2 | XP_006238450.1 | 8 | 1094 | |||
XM_006238389.2 | XP_006238451.1 | 8 | 1631 | |||
XM_006238391.2 | 8 | 1628 | ||||
XM_006238395.2 | XP_006238457.1 | 7 | 1022 | |||
XM_008763848.1 | XP_008762070.1 | 7 | 1337 | |||
XM_008763849.1 | XP_008762071.1 | 7 | 1334 | |||
XM_008763850.1 | XP_008762072.1 | 7 | 1000 | |||
GenBank mRNA | AB098713.1 | - | 8 | 1552 |