Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173310.3 | NP_775432.3 | 1 | 332 | ||
XM_006230430.2 | XP_006230492.1 | 2 | 1899 | |||
XM_006230431.2 | XP_006230493.1 | 2 | 2639 | |||
XM_006230433.2 | XP_006230495.1 | 2 | 2134 | |||
XM_008759868.1 | XP_008758090.1 | 2 | 895 | |||
XM_008759869.1 | XP_008758091.1 | 2 | 2638 | |||
GenBank mRNA | AB086953.1 | - | 1 | 332 | ||
BC085687.1 | - | 1 | 593 | 7189319 |