Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175603.2 | NP_783193.2 | 9, 10 | 1261 | ||
GenBank mRNA | AY184814.1 | - | 8, 9 | 1175 | ||
BC093598.1 | - | 9, 10 | 1261 | 7315896 |