Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem184c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178330.1 | NP_847900.1 | 1, 2 | 587 | ||
XM_006255422.2 | XP_006255484.1 | 2, 3 | 299 | |||
GenBank mRNA | AY228474.1 | - | 1, 2 | 587 | ||
BC089112.1 | - | 1, 2 | 597 | 7316181 |