Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181080.1 | NP_851594.1 | 1 | 320 | ||
XM_006249128.2 | XP_006249190.1 | 2 | 642 | |||
XM_006249129.2 | XP_006249191.1 | 2 | 642 | |||
XM_006249130.2 | XP_006249192.1 | 2 | 642 | |||
GenBank mRNA | AY081950.1 | - | 1 | 320 | ||
BC085684.1 | - | 2 | 480 | 7189609 |