Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019885.3 | NP_063938.1 | 2 | 266 | ||
XM_005264433.4 | XP_005264490.1 | 2 | 230 | |||
GenBank mRNA | AF252297.1 | - | 2 | 238 | ||
AK294933.1 | - | 1 | 236 | |||
AK313433.1 | - | 2 | 241 | |||
BC069443.1 | - | 2 | 238 | 7262360 | ||
BC109205.2 | - | 2 | 238 | 40006245 |