Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181091.2 | NP_851605.1 | 3 | 178 | ||
XM_006228606.2 | XP_006228668.1 | 4 | 476 | |||
XM_006228607.2 | XP_006228669.1 | 4 | 476 | |||
XM_008759100.1 | XP_008757322.1 | 4 | 291 | |||
XM_008759101.1 | XP_008757323.1 | 3 | 220 | |||
XM_008759102.1 | XP_008757324.1 | 3 | 168 | |||
GenBank mRNA | AB007364.1 | - | 3 | 178 |